Sodium octyl sulphate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Octyl sulphate, sodium salt, Octyl sulphuric acid salt
M-Clarity™ quality level = MQ200
O4003-1G
O4003-25G
O4003-5G
SIALO4003-1GEA
2860
CZK
SIALO4003-1G
SIALO4003-25G
SIALO4003-5G
Sodium octyl sulphate, Sigma-Aldrich®
Sodium octyl sulphate
Vzorec:
C₈H₁₇NaO₄S MW: 232,28 g/mol Bod tání: 181…183 °C Hustota: 1,266 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007470 CAS číslo: 142-31-4 EINECS: 205-535-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...