Sodium methyl sulphate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
318183-100G
318183-25G
SIAL318183-100GEA
13180
CZK
SIAL318183-100G
SIAL318183-25G
Sodium methyl sulphate, Sigma-Aldrich®
Sodium methyl sulphate
Vzorec:
CH₃NaO₄S Bod tání: 193 °C |
MDL Number:
MFCD00013457 CAS číslo: 512-42-5 EINECS: 208-142-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...