Tisk…

Diethylsulfát ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L14291.AE L14291.AP
L14291.AEEA 800 CZK
L14291.AE L14291.AP
Diethylsulfát ≥98%
Diethylsulfát

Purity: 98% CAS No.: 64-67-5 Molecular Formula: (CH3CH2O)2SO2 Formula Weight: 154.19 Flash Point: 104°(219°F) Refractive Index: 1.4005 Melting Point: -25° Boiling Point: 208° dec. Beilstein Reference: 1209714 Merck Reference: 14,3130 UN No.: 1594 MDL No.: MFCD00009099

Vzorec: C₄H₁₀O₄S
Bod varu: 208 °C (rozkl.)
Bod tání: –25 °C
Hustota: 1,177
Bod vzplanutí: 104 °C (219 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009099
CAS číslo: 64-67-5
EINECS: 200-589-6
UN: 1594
ADR: 6.1,II
Merck Index: 14,3130

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR