Dibutyl sulphate 95%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Di-n-butylsulphate
L12813.09
L12813.18
L12813.09EA
1860
CZK
L12813.09
L12813.18
Dibutyl sulphate 95%
Dibutyl sulphate
Purity: 95%
CAS No.: 625-22-9
Molecular Formula: C8H18O4S
Formula Weight: 210.29
Refractive Index: 1.4205
Boiling Point: 115-116°/6mm
MDL No.: MFCD00059420
Vzorec:
C₈H₁₈O₄S MW: 210,29 g/mol Bod varu: 115…116 °C (6 mmHg) Hustota: 1,060 Storage Temperature: Ambient |
MDL Number:
MFCD00059420 CAS číslo: 625-22-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...