Glycerin, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol
M-Clarity™ quality level = MQ200
G9012-100ML
G9012-1L
G9012-2L
G9012-500ML
G9012-1GA
SIALG9012-100MLEA
1930
CZK
SIALG9012-100ML
SIALG9012-1L
SIALG9012-2L
SIALG9012-500ML
SIALG9012-1GA
Glycerin, Sigma-Aldrich®
Glycerin
Vzorec:
HOCH₂CH(OH)CH₂OH MW: 92.09 Bod varu: 290 Bod tání: 18.6 Hustota: 1.26 g/cm³ (20 °C) Bod vzplanutí: 177 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004722 CAS číslo: 56-81-5 EINECS: 200-289-5 Merck Index: 13,04497 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...