Glycerin, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol
M-Clarity™ quality level = MQ300
G2289-1L
G2289-4L
G2289-500ML
SIALG2289-1LEA
23220
CZK
SIALG2289-1L
SIALG2289-4L
SIALG2289-500ML
Glycerin, Sigma-Aldrich®
Glycerin
Vzorec:
HOCH₂CH(OH)CH₂OH Bod varu: 290 °C (1013 hPa) Hustota: 1,26 g/cm³ (20 °C) Bod vzplanutí: 177 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004722 CAS číslo: 56-81-5 EINECS: 200-289-5 Merck Index: 13,04497 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...