1,9-Nonanediol, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
1,9-Dihydroxy nonane, Nonane-1,9-diol
M-Clarity™ quality level = MQ100
N29600-25G
N29600-100G
SIALN29600-25GEA
1660
CZK
SIALN29600-25G
SIALN29600-100G
1,9-Nonanediol, Sigma-Aldrich®
1,9-Nonanediol
Vzorec:
C₉H₂₀O₂ MW: 160,26 g/mol Bod varu: 292 °C (1013 hPa) Bod tání: 45…46 °C Hustota: 0,918 g/cm³ (50 °C) Bod vzplanutí: 162 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002991 CAS číslo: 3937-56-2 EINECS: 223-517-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...