Tisk…

Potassium methanolate ≥90%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Potassium methoxide, Potassium methylate

14261.18 14261.30
14261.18EA 1440 CZK
14261.18 14261.30
Potassium methanolate ≥90%
Potassium methanolate

Purity: 90+% CAS No.: 865-33-8 Molecular Formula: CH3KO Formula Weight: 70.14 Physical Form: Powder Sensitivity: Air and Moisture Sensitive UN No.: 3206 MDL No.: MFCD00012416

Vzorec: CH₃KO
MW: 70,13 g/mol
Hustota: 0,85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012416
CAS číslo: 865-33-8
EINECS: 212-736-1
UN: 3206
ADR: 4.2,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR