Tisk…

Potassium-tert-butoxide, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ100

659878-25G 659878-5G
SIAL659878-25GEA 8290 CZK
SIAL659878-25G SIAL659878-5G
Potassium-tert-butoxide, Sigma-Aldrich®
Potassium-tert-butoxide
Vzorec: C₄H₉KO
Bod tání: 256…258 °C
MDL Number: MFCD00012162
CAS číslo: 865-47-4
EINECS: 212-740-3
UN: 3206
ADR: 4.2,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR