Potassium-tert-butoxide, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Terc-butanolát draselný
M-Clarity™ quality level = MQ200
156671-500G
156671-5G
SIAL156671-500GEA
4890
CZK
SIAL156671-500G
SIAL156671-5G
Potassium-tert-butoxide, Sigma-Aldrich®
Potassium-tert-butoxide
Vzorec:
C₄H₉KO Bod tání: 256…258 °C |
MDL Number:
MFCD00012162 CAS číslo: 865-47-4 EINECS: 212-740-3 UN: 3206 ADR: 4.2,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...