Tisk…

1-Heptadecanol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Heptadecan-1-ol, Hexadecyl carbinol, 1-Heptadecanol

M-Clarity™ quality level = MQ100

241695-5G 241695-25G
SIAL241695-5GEA 1890 CZK
SIAL241695-5G SIAL241695-25G
1-Heptadecanol, Sigma-Aldrich®
1-Heptadecanol
Vzorec: H₃C(CH₂)₁₅CH₂OH
MW: 256.47
Bod varu: 308.5
Bod tání: 53…57 °C
Hustota: 0.967 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002822
CAS číslo: 1454-85-9
EINECS: 215-932-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR