1-Heneicosanol, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
H2518-1G
H2518-25MG
SIALH2518-1GEA
31880
CZK
SIALH2518-1G
SIALH2518-25MG
1-Heneicosanol, Sigma-Aldrich®
1-Heneicosanol
Vzorec:
H₃C(CH₂)₁₉CH₂OH MW: 312,58 g/mol Bod varu: 170 °C (1013 hPa) Bod tání: 68…69 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00062834 CAS číslo: 15594-90-8 EINECS: 239-673-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...