Caproic anhydride, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Hexanoic anhydride
M-Clarity™ quality level = MQ200
21580-250ML-F
21580-50ML-F
SIAL21580-250ML-FEA
3030
CZK
SIAL21580-250ML-F
SIAL21580-50ML-F
Caproic anhydride, Sigma-Aldrich®
Caproic anhydride
Vzorec:
C₁₂H₂₂O₃ MW: 214,3 g/mol Bod varu: 246 °C (1013 hPa) Bod tání: –40 °C Hustota: 0,93 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00009509 CAS číslo: 2051-49-2 EINECS: 218-121-4 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...