Tisk…

Anhydrid kyseliny octové ≥98.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent

Dodavatel: VWR Chemicals
Nebezpečí

Synonyms: Acetyl oxide, Acetyl acetate

21390.293EA 2000 CZK
21390.293 21390.330
Anhydrid kyseliny octové ≥98.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Anhydrid kyseliny octové
Vzorec: (CH₃CO)₂O
MW: 102,09 g/mol
Bod varu: 139,9 °C (1013 hPa)
Bod tání: –73 °C
Hustota: 1,08 g/cm³ (20 °C)
Bod vzplanutí: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00008705
CAS číslo: 108-24-7
EINECS: 203-564-8
UN: 1715
ADR: 8 (3),II
REACH: 01-2119486470-36
Merck Index: 13,00057
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f107a66d926eb36bee4d132880765bc7fc2db7c8&vid=1c6b77698bd69c2f562bcda9bf02b8dc694a0d16&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Assay Min. 98.0 %
IR Spectrum Passes test
Substances reducing KMnO4 Passes test ACS
Boiling point 136 - 142 °C
Colouration Max. 20 APHA
Substances coloured by H2SO4 Max. 100 APHA
Evaporation residue Max. 30 ppm
Heavy metals (by ICP-MS) Max. 2 ppm
Substances reducing Cr6+ (as O) Max. 0.03 %
Cl (Chloride) Max. 2 ppm
PO4 (Phosphate) Max. 5 ppm
SO4 (Sulphate) Max. 5 ppm
Cd (Cadmium) Max. 0.05 ppm
Cu (Copper) Max. 0.05 ppm
Fe (Iron) Max. 2 ppm
Ni (Nickel) Max. 0.05 ppm
Pb (Lead) Max. 0.05 ppm
Zn (Zinc) Max. 0.1 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR