Glutaric anhydride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Dihydro-2H-pyran-2,6(3H)-dione, Dihydro-2H-pyran-2,6(3H)-dione
M-Clarity™ quality level = MQ200
G3806-100G
G3806-500G
G3806-5G
SIALG3806-100GEA
2590
CZK
SIALG3806-100G
SIALG3806-500G
SIALG3806-5G
Glutaric anhydride, Sigma-Aldrich®
Glutaric anhydride
Vzorec:
C₅H₆O₃ MW: 114.1 Bod varu: 287 Bod tání: 52…55 °C Hustota: 1.411 g/cm³ (20 °C) Bod vzplanutí: 174 Storage Temperature: Ambient |
MDL Number:
MFCD00006679 CAS číslo: 108-55-4 EINECS: 203-593-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...