4-Methoxy-1-butanol ≥98%
Dodavatel: Thermo Fisher Scientific
Varování
|
H50339.03
H50339.06
H50339.03EA
1460
CZK
H50339.03
H50339.06
4-Methoxy-1-butanol ≥98%
4-Methoxy-1-butanol
Purity: 98+%
CAS No.: 111-32-0
Molecular Formula: C5H12O2
Formula Weight: 104.15
Flash Point: 73°(163°F)
Odour: Mild
Boiling Point: 66°/7mm
MDL No.: MFCD00040437
Vzorec:
C₅H₁₂O₂ Bod varu: 66 °C (7 mmHg) Hustota: 0,93 Bod vzplanutí: 73 °C (163 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040437 CAS číslo: 111-32-0 EINECS: 203-858-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...