Tisk…

Kyselina 2-(cyklohexylamino)ethansulfonová, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: N-Cyclohexyltaurine, CHES

M-Clarity™ quality level = MQ200

C8210-100G C8210-25G
SIALC8210-100GEA 8030 CZK
SIALC8210-100G SIALC8210-25G
Kyselina 2-(cyklohexylamino)ethansulfonová, Sigma-Aldrich®
Kyselina 2-(cyklohexylamino)ethansulfonová
Vzorec: C₈H₁₇NO₃S
MW: 207,29 g/mol
Bod tání: 320 °C
Storage Temperature: Ambient
MDL Number: MFCD00003835
CAS číslo: 103-47-9
EINECS: 203-115-6
Merck Index: 13,02063

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR