Kyselina 2-(cyklohexylamino)ethansulfonová, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
N-Cyclohexyltaurine, CHES
29311-10G
29311-50G
SIAL29311-10GEA
2560
CZK
SIAL29311-10G
SIAL29311-50G
Kyselina 2-(cyklohexylamino)ethansulfonová, Sigma-Aldrich®
Kyselina 2-(cyklohexylamino)ethansulfonová
Vzorec:
C₈H₁₇NO₃S MW: 207.29 Bod tání: 320 Storage Temperature: Ambient |
MDL Number:
MFCD00003835 CAS číslo: 103-47-9 EINECS: 203-115-6 Merck Index: 13,02063 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...