Tisk…

Kyselina 2-(cyklohexylamino)ethansulfonová ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: N-Cyclohexyltaurine, CHES

A18047.14 A18047.22 A18047.36 A18047.99
A18047.14EA 1620 CZK
A18047.14 A18047.22 A18047.36 A18047.99
Kyselina 2-(cyklohexylamino)ethansulfonová ≥99%
Kyselina 2-(cyklohexylamino)ethansulfonová

Purity: 99% CAS No.: 103-47-9 Molecular Formula: C8H17NO3S Formula Weight: 207.29 Melting Point: 300° Beilstein Reference: 2967601 Merck Reference: 14,2055 MDL No.: MFCD00003835


Zwitterionic buffer useful in pH range 8.6 to 10.0

Vzorec: C₈H₁₇NO₃S
MW: 207.29
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00003835
CAS číslo: 103-47-9
EINECS: 203-115-6
Merck Index: 13,02063

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR