Tisk…

Bis-trispropane, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1,3-Bis(tris(hydroxymethyl)methylamino)propane, 1,3-Bis[tris(hydroxymethyl)amino]propane, 2,2'-(Propane-1,3-diylbis(azanediyl))bis(2-(hydroxymethyl)propane-1,3-diol), BIS-TRIS propane, 2,2'-(Propane-1,3-diyldiimino)bis[2-(hydroxymethyl)propane-1,3-diol]

M-Clarity™ quality level = MQ400

B4679-100G B4679-25G B4679-500G
SIALB4679-100GEA 13280 CZK
SIALB4679-100G SIALB4679-25G SIALB4679-500G
Bis-trispropane, Sigma-Aldrich®
Bis-trispropane
Vzorec: C₁₁H₂₆N₂O₆
MW: 282,34 g/mol
MDL Number: MFCD00004689
CAS číslo: 64431-96-5
EINECS: 264-899-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR