Spermidine, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
S2626-1G
S2626-25G
S2626-5G
SIALS2626-1GEA
1890
CZK
SIALS2626-1G
SIALS2626-25G
SIALS2626-5G
Spermidine, Sigma-Aldrich®
Spermidine
Vzorec:
C₇H₁₉N₃ Bod varu: 246 °C (1013 hPa) Bod tání: 20…25 °C Hustota: 0,925 g/cm³ (20 °C) |
MDL Number:
MFCD00008229 CAS číslo: 124-20-9 EINECS: 204-689-0 Merck Index: 12,08893 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...