Tisk…

Spermidine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: N1-(3-Aminopropyl)butane-1,4-diamine, N-(3-Aminopropyl)-1,4-diaminobutane, 4-Azaoctamethylenediamine

M-Clarity™ quality level = MQ200

S0266-1G S0266-25G S0266-5G
SIALS0266-1GEA 2480 CZK
SIALS0266-1G SIALS0266-25G SIALS0266-5G
Spermidine, Sigma-Aldrich®
Spermidine
Vzorec: C₇H₁₉N₃
MW: 145,25 g/mol
Bod varu: 246 °C (1013 hPa)
Bod tání: 20…25 °C
Hustota: 0,925 g/cm³ (20 °C)
MDL Number: MFCD00008229
CAS číslo: 124-20-9
EINECS: 204-689-0
Merck Index: 12,08893

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR