Spermidine ≥99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A19096.03
A19096.06
A19096.14
A19096.22
A19096.03EA
1370
CZK
A19096.03
A19096.06
A19096.14
A19096.22
Spermidine ≥99%
Spermidine
Purity: 99%
CAS No.: 124-20-9
Molecular Formula: H2N(CH2)3NH(CH2)4NH2
Formula Weight: 145.25
Flash Point:
Vzorec:
C₇H₁₉N₃ Bod varu: 128…130 °C (14 mmHg) Bod tání: 22…25 °C Hustota: 0,925 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008229 CAS číslo: 124-20-9 EINECS: 204-689-0 ADR: 8,III Merck Index: 14,8742 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...