Tisk…

N-Methylethylenediamine 95%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 2-Aminoethylmethylamine, N1-Methylethane-1,2-diamine

155560050 155560250 155561000
ACRO155560050EA 2340 CZK
ACRO155560050 ACRO155560250 ACRO155561000
N-Methylethylenediamine 95%
N-Methylethylenediamine
Vzorec: C₃H₁₀N₂
MW: 74,13 g/mol
Bod varu: 114…116 °C (760 mmHg)
Hustota: 0,84 g/cm³
Bod vzplanutí: 41 °C
Storage Temperature: Ambient
MDL Number: MFCD00008165
CAS číslo: 109-81-9
EINECS: 203-707-4
UN: 2734
ADR: 8,II

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4390 to 1.4420 (20°C, 589 nm)
Titration with HClO4 ≥94.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR