Tisk…

Dioctylamine, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ200

D201146-100G D201146-25G
SIALD201146-100GEA 3660 CZK
SIALD201146-100G SIALD201146-25G
Dioctylamine, Sigma-Aldrich®
Dioctylamine
Vzorec: C₁₆H₃₅N
Bod varu: 298 °C (1013 hPa)
Hustota: 0,8026 g/cm³ (20 °C)
MDL Number: MFCD00009557
CAS číslo: 1120-48-5
EINECS: 214-311-6
UN: 2735
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR