Tisk…

N,N'-Dimethylethylendiamine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: N1,N2-Dimethylethane-1,2-diamine, sym. Dimethylethylendiamine, N,N'-Dimethylethylenediamine

M-Clarity™ quality level = MQ200

D157805-25G D157805-100G D157805-5G
SIALD157805-25GEA 8880 CZK
SIALD157805-25G SIALD157805-100G SIALD157805-5G
N,N'-Dimethylethylendiamine, Sigma-Aldrich®
N,N'-Dimethylethylendiamine
Vzorec: C₄H₁₂N₂
MW: 88,15 g/mol
Bod varu: 119 °C (1013 hPa)
Hustota: 0,82 g/cm³ (20 °C)
MDL Number: MFCD00008290
CAS číslo: 110-70-3
EINECS: 203-793-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR