Tisk…

N,N'-Dimethylethylendiamine 95%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: N1,N2-Dimethylethane-1,2-diamine, sym. Dimethylethylendiamine, N,N'-Dimethylethylenediamine

L02204.06 L02204.14
L02204.06EA 1590 CZK
L02204.06 L02204.14
N,N'-Dimethylethylendiamine 95%
N,N'-Dimethylethylendiamine

Purity: 95% CAS No.: 110-70-3 Molecular Formula: C4H12N2 Formula Weight: 88.15 Flash Point: 28°(82°F) Refractive Index: 1.4300 Odour: Amine-like Boiling Point: 118-119° Beilstein Reference: 878142 UN No.: 2734 MDL No.: MFCD00008290

Vzorec: C₄H₁₂N₂
MW: 88.15
Bod varu: 118…119 °C
Hustota: 0.82
Bod vzplanutí: 28 °C (82 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008290
CAS číslo: 110-70-3
EINECS: 203-793-3
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR