Tisk…

1,7-Diaminoheptane 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L02853.06 L02853.14
L02853.06EA 1240 CZK
L02853.06 L02853.14
1,7-Diaminoheptane 98%
1,7-Diaminoheptane

Purity: 98% CAS No.: 646-19-5 Molecular Formula: C7H18N2 Formula Weight: 130.24 Flash Point: 87°(188°F) Refractive Index: 1.4590 Odour: Amine-like Melting Point: 26-30° Boiling Point: 223-225° Beilstein Reference: 1734159 UN No.: 3259 MDL No.: MFCD00008246

Vzorec: C₇H₁₈N₂
Bod varu: 223…225 °C
Bod tání: 26…30 °C
Hustota: 0,860
Bod vzplanutí: 87 °C (188 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008246
CAS číslo: 646-19-5
EINECS: 211-468-2
UN: 3259
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR