Tisk…

1,5-Diaminopentane, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: 1,5-Pentanediamine, Pentamethylendiamine, Cadaverine, Cadavarine

M-Clarity™ quality level = MQ200

33211-10ML-F 33211-50ML-F
SIAL33211-10ML-FEA 6020 CZK
SIAL33211-10ML-F SIAL33211-50ML-F
1,5-Diaminopentane, Sigma-Aldrich®
1,5-Diaminopentane
Vzorec: C₅H₁₄N₂
MW: 102.18
Bod varu: 178
Hustota: 0.9174 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008239
CAS číslo: 462-94-2
EINECS: 207-329-0
UN: 2735
ADR: 8 III
Merck Index: 12,01645

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR