1,4-Diaminobutane, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Putrescine, 1,4-Butanediamine, Tetramethylendiamine
M-Clarity™ quality level = MQ200
D13208-100G
D13208-25G
D13208-500G
SIALD13208-100GEA
3210
CZK
SIALD13208-100G
SIALD13208-25G
SIALD13208-500G
1,4-Diaminobutane, Sigma-Aldrich®
1,4-Diaminobutane
Vzorec:
C₄H₁₂N₂ MW: 88.15 Bod varu: 158 Bod tání: 25…28 °C Hustota: 0.88 g/cm³ (20 °C) Bod vzplanutí: 63 Storage Temperature: Ambient |
MDL Number:
MFCD00008235 CAS číslo: 110-60-1 EINECS: 203-782-3 UN: 2928 ADR: 6.1 II Merck Index: 12,08134 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...