1,3-Diaminopropan 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
1,3-Propanediamine, Trimethylenediamine
A11932.0F
A11932.AE
A11932.AP
A11932.0FEA
6800
CZK
A11932.0F
A11932.AE
A11932.AP
1,3-Diaminopropan 98%
1,3-Diaminopropane
Purity: 98%
CAS No.: 109-76-2
Molecular Formula: H2N(CH2)3NH2
Formula Weight: 74.13
Flash Point: 48°(118°F)
Refractive Index: 1.4573
Odour: Amine-like
pH:
Vzorec:
C₃H₁₀N₂ MW: 74,13 g/mol Bod varu: 134…136 °C Bod tání: –12 °C Hustota: 0,888 Bod vzplanutí: 48 °C (118 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008228 CAS číslo: 109-76-2 EINECS: 203-702-7 UN: 2734 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...