1,2-Propylendiamin, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
1,2-Diaminopropane
M-Clarity™ quality level = MQ200
117498-100G
117498-500G
117498-2KG
SIAL117498-100GEA
653
CZK
SIAL117498-100G
SIAL117498-500G
SIAL117498-2KG
1,2-Propylendiamin, Sigma-Aldrich®
1,2-Propylendiamin
Vzorec:
C₃H₁₀N₂ MW: 74.13 Bod varu: 120.5 Bod tání: –37 °C Hustota: 0.8584 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008089 CAS číslo: 78-90-0 EINECS: 201-155-9 UN: 2258 ADR: 8 II Merck Index: 12,08037 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...