1,12-Diaminododecane, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
N12N, Dodecamethylendiamine, 1,12-Dodecanediamine
M-Clarity™ quality level = MQ200
D16401-25G
D16401-5G
SIALD16401-25GEA
3670
CZK
SIALD16401-25G
SIALD16401-5G
1,12-Diaminododecane, Sigma-Aldrich®
1,12-Diaminododecane
Vzorec:
C₁₂H₂₈N₂ MW: 200,37 g/mol Bod varu: 275 °C (1013 hPa) Storage Temperature: Ambient |
MDL Number:
MFCD00008155 CAS číslo: 2783-17-7 EINECS: 220-489-6 UN: 3259 ADR: 8,III |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...