Taurin, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-Aminoethanesulphonic acid
86329-25G
86329-500G
SIAL86329-25GEA
4510
CZK
SIAL86329-25G
SIAL86329-500G
Taurin, Sigma-Aldrich®
Taurin
Vzorec:
C₂H₇NO₃S MW: 125,15 g/mol Bod tání: 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008197 CAS číslo: 107-35-7 EINECS: 203-483-8 Merck Index: 13,09163 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...