Tris-(hydroxymethyl)aminomethan, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
M-Clarity™ quality level = MQ400
T4661-100G
T4661-1KG
T4661-5KG
SIALT4661-100GEA
1920
CZK
SIALT4661-100G
SIALT4661-1KG
SIALT4661-5KG
Tris-(hydroxymethyl)aminomethan, Sigma-Aldrich®
Tris-(hydroxymethyl)aminomethan
Vzorec:
NH₂C(CH₂OH)₃ Bod varu: 357 °C (1013 hPa) Bod tání: 169…172 °C Hustota: 1,328 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004679 CAS číslo: 77-86-1 EINECS: 201-064-4 Merck Index: 13,09842 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...