4-Aminobutan-2-ol 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
4-Amino-2-butanol
330230010
330230050
ACRO330230010EA
6140
CZK
ACRO330230010
ACRO330230050
4-Aminobutan-2-ol 98%
4-Aminobutan-2-ol
Vzorec:
C₄H₁₁NO MW: 89,14 g/mol Bod varu: 103…104 °C (34 mmHg) Hustota: 0,93 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00216936 CAS číslo: 39884-48-5 EINECS: 254-673-2 UN: 2735 ADR: 8,III |
Výsledky specifikačního testu
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4518 to 1.4538 (20°C, 589 nm) |
GC | ≥97.5 % |
Titration with HClO4 | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...