4-Aminobutan-1-ol 98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
4-Hydroxybutylamine, 4-Amino-1-butanol
A12680.03
A12680.06
A12680.14
A12680.03EA
951
CZK
A12680.03
A12680.06
A12680.14
4-Aminobutan-1-ol 98%
4-Aminobutan-1-ol
Purity: 98%
CAS No.: 13325-10-5
Molecular Formula: C4H11NO
Formula Weight: 89.14
Flash Point: 107°(224°F)
Refractive Index: 1.4610
Odour: Amine-like
Melting Point: 16-18°
Boiling Point: 206°
Beilstein Reference: 1731411
UN No.: 2735
MDL No.: MFCD00008230
Vzorec:
C₄H₁₁NO Bod varu: 206 °C Bod tání: 16…18 °C Hustota: 0,967 Bod vzplanutí: 107 °C (224 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008230 CAS číslo: 13325-10-5 EINECS: 236-364-4 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...