Tetradecylamine, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
1-Tetradecylamine, 1-Aminotetradecane, n-Tetradecylamine, Tetradecan-1-amine
M-Clarity™ quality level = MQ100
T10200-25G
T10200-5G
SIALT10200-25GEA
4850
CZK
SIALT10200-25G
SIALT10200-5G
Tetradecylamine, Sigma-Aldrich®
Tetradecylamine
Vzorec:
H₃C(CH₂)₁₂CH₂NH₂ MW: 213,41 g/mol Bod varu: 291 °C (1013 hPa) Bod tání: 40 °C Hustota: 0,8571 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008157 CAS číslo: 2016-42-4 EINECS: 217-950-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...