Ethyl isocyanate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
E33300-25G
E33300-5G
SIALE33300-25GEA
3310
CZK
SIALE33300-25G
SIALE33300-5G
Ethyl isocyanate, Sigma-Aldrich®
Ethyl isocyanate
Vzorec:
C₃H₅NO Bod varu: 60 °C (1013 hPa) Bod tání: < –50 °C Hustota: 0,9 g/cm³ (20 °C) Storage Temperature: Ambient |
CAS číslo:
109-90-0 EINECS: 203-717-9 UN: 2481 ADR: 3,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...