Octadecyl isocyanate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
O1807-100G
O1807-500G
SIALO1807-100GEA
2410
CZK
SIALO1807-100G
SIALO1807-500G
Octadecyl isocyanate, Sigma-Aldrich®
Octadecyl isocyanate
Vzorec:
C₁₉H₃₇NO Bod varu: 140…141 °C (0,4 mmHg) Bod tání: 15…16 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00002041 CAS číslo: 112-96-9 EINECS: 204-019-7 UN: 2206 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...