Tisk…

1,4-Diisocyanatobutane, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Tetramethylene diisocyanate

M-Clarity™ quality level = MQ100

69006-1ML 69006-5ML
SIAL69006-1MLEA 0 CZK
SIAL69006-1ML SIAL69006-5ML
1,4-Diisocyanatobutane, Sigma-Aldrich®
1,4-Diisocyanatobutane
Vzorec: C₆H₈N₂O₂
MW: 140,14 g/mol
Bod varu: 108…110 °C (15 mmHg)
Bod tání: 185…187 °C
Hustota: 1,1 g/cm³ (20 °C)
MDL Number: MFCD00075559
CAS číslo: 4538-37-8
UN: 3334
ADR: 9,

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR