Tisk…

1,2-Bis(dimethylamino)methane, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: N,N,N',N'-Tetramethylmethylenediamine, N,N,N',N'-Tetramethyldiaminomethane

M-Clarity™ quality level = MQ200

T21407-100G T21407-25G
SIALT21407-100GEA 5310 CZK
SIALT21407-100G SIALT21407-25G
1,2-Bis(dimethylamino)methane, Sigma-Aldrich®
1,2-Bis(dimethylamino)methane
Vzorec: C₅H₁₄N₂
Bod varu: ∼85 °C (1013 hPa)
Bod tání: –55 °C
Hustota: 0,7494 g/cm³ (20 °C)
Bod vzplanutí: –11 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008328
CAS číslo: 51-80-9
EINECS: 200-124-7
UN: 2733
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR