Tisk…

N,N,N'-Trimethylethylenediamine ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

31785.03 31785.06 31785.14
31785.03EA 1040 CZK
31785.03 31785.06 31785.14
N,N,N'-Trimethylethylenediamine ≥98%
N,N,N'-Trimethylethylenediamine

Purity: 98% CAS No.: 142-25-6 Molecular Formula: C5H14N2 Formula Weight: 102.18 Flash Point: 9°(49°F) Physical Form: Liquid Density: 0.804 Refractive Index: 1.4190 Odour: Amine-like Boiling Point: 116-118° UN No.: 2733 MDL No.: MFCD00014874

Vzorec: C₅H₁₄N₂
Bod varu: 116…118 °C
Hustota: 0,804
Bod vzplanutí: 9 °C (49 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014874
CAS číslo: 142-25-6
EINECS: 205-529-2
UN: 2733
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR