N,N-Dimethyloctylamine 95%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
N,N-Dimethyloctan-1-amine
A12546.09
A12546.18
A12546.30
A12546.09EA
1060
CZK
A12546.09
A12546.18
A12546.30
N,N-Dimethyloctylamine 95%
N,N-Dimethyloctylamine
Purity: 95%
CAS No.: 7378-99-6
Molecular Formula: CH3(CH2)7N(CH3)2
Formula Weight: 157.30
Flash Point: 65°(149°F)
Refractive Index: 1.4243
Melting Point: -57°
Boiling Point: 195°
Beilstein Reference: 1698081
UN No.: 2922
MDL No.: MFCD00009558
Vzorec:
C₁₀H₂₃N MW: 157,3 g/mol Bod varu: 195 °C Bod tání: –57 °C Hustota: 0,765 Bod vzplanutí: 65 °C (149 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009558 CAS číslo: 7378-99-6 EINECS: 230-939-3 UN: 2922 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...