Tisk…

N,N-Dimethyldodecylamine, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: N,n-dimethyldodecylamine, Dodecyldimethylamine

M-Clarity™ quality level = MQ200

284386-1L 284386-250ML
SIAL284386-1LEA 17160 CZK
SIAL284386-1L SIAL284386-250ML
N,N-Dimethyldodecylamine, Sigma-Aldrich®
N,N-Dimethyldodecylamine
Vzorec: C₁₄H₃₁N
MW: 213,41 g/mol
Bod varu: 265 °C (1013 hPa)
Bod tání: –20 °C
Hustota: 0,787 g/cm³ (20 °C)
MDL Number: MFCD00008970
CAS číslo: 112-18-5
EINECS: 203-943-8
UN: 2735
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR