Tisk…

Triethanolamin, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Tris-(2-hydroxyethyl)amine, 2,2',2''-Nitrilotriethanol

M-Clarity™ quality level = MQ200

33729-1L 33729-30KG
SIAL33729-1LEA 3940 CZK
SIAL33729-1L SIAL33729-30KG
Triethanolamin, Sigma-Aldrich®
Triethanolamin
Vzorec: C₆H₁₅NO₃
Bod varu: 335,4 °C (1013 hPa)
Hustota: 1,13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002855
CAS číslo: 102-71-6
EINECS: 203-049-8
Merck Index: 13,09739
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c181d3e80375aec7c34e17766247484e1c3d3257&vid=40c3d3d0c0162ac22208cadb8d5254a7cf0b340a&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR