Tisk…

N,N-Dimethylformamiddimethylacetal, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: 1,1-Dimethoxytrimethylamine, Esterate M, N,N-Dimethylformamide Dimethyl Acetal, 1,1-Dimethoxy-N,N-dimethylmethylamine, DMF-DMA

M-Clarity™ quality level = MQ200

140732-100ML 140732-25ML 140732-500ML
SIAL140732-100MLEA 6240 CZK
SIAL140732-100ML SIAL140732-25ML SIAL140732-500ML
N,N-Dimethylformamiddimethylacetal, Sigma-Aldrich®
N,N-Dimethylformamiddimethylacetal
Vzorec: C₅H₁₃NO₂
MW: 119,16 g/mol
Bod varu: 102 °C (1013 hPa)
Hustota: 0,893 g/cm³ (20 °C)
Bod vzplanutí: 6 °C
MDL Number: MFCD00008482
CAS číslo: 4637-24-5
EINECS: 225-063-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR