Tisk…

Kyselina jodoctová ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

A12410.14 A12410.22
A12410.14EA 2550 CZK
A12410.14 A12410.22
Kyselina jodoctová ≥97%
Kyselina jodoctová

Purity: 97+% CAS No.: 64-69-7 Molecular Formula: ICH2CO2H Formula Weight: 185.95 Storage Temperature: 2°C to 8°C Odour: Pungent Boiling Point: 208° dec. Beilstein Reference: 1739079 Merck Reference: 14,5025 UN No.: 2923 MDL No.: MFCD00002685

Vzorec: C₂H₃IO₂
MW: 185,95 g/mol
Bod varu: 208 °C (rozkl.)
Bod tání: 79…82 °C
Hustota: 2,2694 g/cm³ (85 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002685
CAS číslo: 64-69-7
EINECS: 200-590-1
UN: 2923
ADR: 8,II
Merck Index: 14,5025

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR