Tisk…

Iodoacetamide, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: 2-Iodoacetamide

M-Clarity™ quality level = MQ300

I6125-25G I6125-100G I6125-10G I6125-1KG I6125-5G
SIALI6125-25GEA 6220 CZK
SIALI6125-25G SIALI6125-100G SIALI6125-10G SIALI6125-1KG SIALI6125-5G
Iodoacetamide, Sigma-Aldrich®
Iodoacetamide
Vzorec: ICH₂CONH₂
Bod varu: 266 °C (1013 hPa)
Bod tání: 91…94 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008028
CAS číslo: 144-48-9
EINECS: 205-630-1
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR