2-Iodobutane, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-Butyl iodide, sec-Butyl iodide
M-Clarity™ quality level = MQ100
244325-100G
244325-5G
SIAL244325-100GEA
2870
CZK
SIAL244325-100G
SIAL244325-5G
2-Iodobutane, Sigma-Aldrich®
2-Iodobutane
Vzorec:
C₄H₉I MW: 184,02 g/mol Bod varu: 119 °C (1013 hPa) Bod tání: –104 °C Hustota: 1,6263 g/cm³ (0 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001072 CAS číslo: 513-48-4 EINECS: 208-163-1 UN: 2390 ADR: 3,II Merck Index: 13,01573 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...